Thermo Scientific Chemicals

2-Chloromethyl-4-methoxy-3,5-dimethylpyridine hydrochloride, 98%, Thermo Scientific Chemicals

Catalog number: L19453.14
25 g, Each
Thermo Scientific Chemicals

2-Chloromethyl-4-methoxy-3,5-dimethylpyridine hydrochloride, 98%, Thermo Scientific Chemicals

Catalog number: L19453.14
25 g, Each
Quantity
Catalog number: L19453.14
also known as L19453-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
86604-75-3
IUPAC Name
2-(chloromethyl)-4-methoxy-3,5-dimethylpyridine hydrochloride
Molecular Formula
C9H13Cl2NO
InChI Key
LCJDHJOUOJSJGS-UHFFFAOYSA-N
SMILES
Cl.COC1=C(C)C(CCl)=NC=C1C
Form
Powder or crystalline powder
Appearance (Color)
White to pale cream
Assay (Titration ex Chloride)
≥96.0 to ≤104.0%

Description

2-Chloromethyl-4-methoxy-3,5-dimethylpyridine hydrochloride is used as pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Chloromethyl-4-methoxy-3,5-dimethylpyridine hydrochloride is used as pharmaceutical intermediates.

Solubility
Soluble in water.

Notes
Incompatible with oxidizing agents. Store in a cool, dry conditions in well sealed container.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text