N-Boc-2-aminoisobutyric acid, 98+%, Thermo Scientific Chemicals
N-Boc-2-aminoisobutyric acid, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

N-Boc-2-aminoisobutyric acid, 98+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Catalog number L19484.06
also known as L19484-06
Price (USD)/ Each
43.60
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
43.60
Add to cart
N-Boc-2-aminoisobutyric acid, 98+%, Thermo Scientific Chemicals
Catalog numberL19484.06
Price (USD)/ Each
43.60
-
Add to cart
Chemical Identifiers
CAS30992-29-1
IUPAC Name2-{[(tert-butoxy)carbonyl]amino}-2-methylpropanoate
Molecular FormulaC9H16NO4
InChI KeyMFNXWZGIFWJHMI-UHFFFAOYSA-M
SMILESCC(C)(C)OC(=O)NC(C)(C)C([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
FormCrystals or powder or crystalline powder
Assay (Aqueous acid-base Titration)≥98.0 to ≤102.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only