N-Boc-ethylenediamine hydrochloride, 98%, Thermo Scientific Chemicals
N-Boc-ethylenediamine hydrochloride, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

N-Boc-ethylenediamine hydrochloride, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number L19485.03
also known as L19485-03
Price (USD)/ Each
108.00
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)/ Each
108.00
Add to cart
N-Boc-ethylenediamine hydrochloride, 98%, Thermo Scientific Chemicals
Catalog numberL19485.03
Price (USD)/ Each
108.00
-
Add to cart
Chemical Identifiers
CAS79513-35-2
IUPAC Namehydrogen tert-butyl N-(2-aminoethyl)carbamate chloride
Molecular FormulaC7H17ClN2O2
InChI KeyXUHJJLCKXZTUJN-UHFFFAOYSA-N
SMILES[H+].[Cl-].CC(C)(C)OC(=O)NCCN
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream
Assay (Titration ex Chloride)≥97.5 to ≤102.5%
FormCrystals or powder or crystalline powder
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only