N-Boc-1,6-diaminohexane hydrochloride, 98%, Thermo Scientific Chemicals
N-Boc-1,6-diaminohexane hydrochloride, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

N-Boc-1,6-diaminohexane hydrochloride, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Catalog number L19487.06
also known as L19487-06
Price (USD)/ Each
204.00
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
204.00
Add to cart
N-Boc-1,6-diaminohexane hydrochloride, 98%, Thermo Scientific Chemicals
Catalog numberL19487.06
Price (USD)/ Each
204.00
-
Add to cart
Chemical Identifiers
CAS65915-94-8
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream or pale yellow
Assay (Non-aqueous acid-base Titration)≥97.5 to ≤102.5%
FormCrystals or powder or crystalline powder
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only