N-Boc-(R)-(-)-thiazolidine-4-carboxylic acid, 98%, Thermo Scientific Chemicals
N-Boc-(R)-(-)-thiazolidine-4-carboxylic acid, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

N-Boc-(R)-(-)-thiazolidine-4-carboxylic acid, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Catalog number L19488.06
also known as L19488-06
Price (USD)/ Each
138.00
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
138.00
Add to cart
N-Boc-(R)-(-)-thiazolidine-4-carboxylic acid, 98%, Thermo Scientific Chemicals
Catalog numberL19488.06
Price (USD)/ Each
138.00
-
Add to cart
Chemical Identifiers
CAS51077-16-8
IUPAC Name3-[(tert-butoxy)carbonyl]-1,3-thiazolidine-4-carboxylic acid
Molecular FormulaC9H15NO4S
InChI KeyFJWNZTPXVSWUKF-UHFFFAOYNA-N
SMILESCC(C)(C)OC(=O)N1CSCC1C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (Aqueous acid-base Titration)≥97.5 to ≤102.5%
Appearance (Color)White
FormCrystals or powder or crystalline powder
N-Boc-(R)-(-)-thiazolidine-4-carboxylic acid is used as pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Boc-(R)-(-)-thiazolidine-4-carboxylic acid is used as pharmaceutical intermediates.

Solubility
Insoluble in water.

Notes
Store at 4°C. Store in a cool, dry conditions in well sealed container. Store away from oxidizing agents.
RUO – Research Use Only

General References:

  1. Takuya Seko; Masashi Kato; Hiroshi Kohno; Shizuka Ono; Kazuya Hashimura; Hideyuki Takimizu; Katsuhiko Nakai; Hitoshi Maegawa; Nobuo Katsube; Masaaki Toda. Structure-Activity Study of l-Cysteine-Based N-Type Calcium Channel Blockers: Optimization of N- and C-Terminal Substituents. Bioorganic & Medicinal Chemistry Letters.2002, 12, 915-918.