Thermo Scientific Chemicals

(S)-(-)-3-Hydroxypyrrolidine, 99%, ee 99%, Thermo Scientific Chemicals

Catalog number: L19498.06
5 g, Each
Thermo Scientific Chemicals

(S)-(-)-3-Hydroxypyrrolidine, 99%, ee 99%, Thermo Scientific Chemicals

Catalog number: L19498.06
5 g, Each
Quantity
Catalog number: L19498.06
also known as L19498-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
100243-39-8
Appearance (Color)
Colorless to white to yellow
Form
Crystals or powder or crystalline powder or lumps or fused solid or clear liquid as melt
Assay (GC)
≥97.0%

Description

(S)-(-)-3-Hydroxypyrrolidine is an important raw material and intermediate used in organic synthesis, pharmaceuticals agrochemicals and dyestuff fields.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(S)-(-)-3-Hydroxypyrrolidine is an important raw material and intermediate used in organic synthesis, pharmaceuticals agrochemicals and dyestuff fields.

Solubility
Fully miscible in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents. It is hygroscopic and sensitive to air.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text