Thermo Scientific Chemicals

(R)-(+)-3-Hydroxypyrrolidine, 99%, ee 99%, Thermo Scientific Chemicals

Catalog number: L19499.06
5 g, Each
Thermo Scientific Chemicals

(R)-(+)-3-Hydroxypyrrolidine, 99%, ee 99%, Thermo Scientific Chemicals

Catalog number: L19499.06
5 g, Each
Quantity
Catalog number: L19499.06
also known as L19499-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
2799-21-5
IUPAC Name
(3R)-3-hydroxypyrrolidin-1-ium
Molecular Formula
C4H10NO
InChI Key
JHHZLHWJQPUNKB-SCSAIBSYSA-O
SMILES
O[C@@H]1CC[NH2+]C1
Appearance (Color)
Clear, colourless to yellow
Form
liquid as melt
Assay (GC)
>98.5%
Refractive Index
1.4880-1.4920
Optical Rotation
6.5 ?1? (c=3.5 in methanol)

Description

(R)-(+)-3-Hydroxypyrrolidine is used in the enantioselective addition of diethylzinc to aromatic aldehydes leading to optically active secondary alcohols. Also used to prepare-κ-opioid receptor agonists.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(R)-(+)-3-Hydroxypyrrolidine is used in the enantioselective addition of diethylzinc to aromatic aldehydes leading to optically active secondary alcohols. Also used to prepare-κ-opioid receptor agonists.

Solubility
Soluble in water and methanol.

Notes
Store under inert gas. It is sensitive to moisture. Store in a cool, dry conditions in well sealed containers. It is hygroscopic and sensitive to air. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text