Thermo Scientific Chemicals

6-Bromopyridine-2-carboxaldehyde, 97%, Thermo Scientific Chemicals

Catalog number: L19518.06
5 g, Each
Thermo Scientific Chemicals

6-Bromopyridine-2-carboxaldehyde, 97%, Thermo Scientific Chemicals

Catalog number: L19518.06
5 g, Each
Quantity
Catalog number: L19518.06
also known as L19518-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
34160-40-2
IUPAC Name
6-bromopyridine-2-carbaldehyde
Molecular Formula
C6H4BrNO
InChI Key
QWFHFNGMCPMOCD-UHFFFAOYSA-N
SMILES
BrC1=CC=CC(C=O)=N1
Appearance (Color)
White to cream to pale brown
Form
Crystals or powder or crystalline powder
Assay (GC)
≥96.0%
Identification (FTIR)
Conforms
Melting Point
75.0-85.0°C

Description

6-Bromopyridine-2-carboxaldehyde is used in the study of a rhodium-catalyzed, reductive aldol coupling with divinyl ketones leading to syn ß-hydroxyenones. It is also used as a building block for Tris[(pyridyl)methyl]amine ligands.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
6-Bromopyridine-2-carboxaldehyde is used in the study of a rhodium-catalyzed, reductive aldol coupling with divinyl ketones leading to syn ß-hydroxyenones. It is also used as a building block for Tris[(pyridyl)methyl]amine ligands.

Solubility
Soluble in dichloromethane, ether, ethyl acetate and methanol. Insoluble in water.

Notes
Store under inert gas. It is sensitive to air. Store in a cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text