Thermo Scientific Chemicals

3-Bromo-4-nitropyridine N-oxide, 98+%, Thermo Scientific Chemicals

Catalog number: L19524.06
5 g, Each
Thermo Scientific Chemicals

3-Bromo-4-nitropyridine N-oxide, 98+%, Thermo Scientific Chemicals

Catalog number: L19524.06
5 g, Each
Quantity
Catalog number: L19524.06
also known as L19524-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1678-49-5
IUPAC Name
3-bromo-4-nitropyridin-1-ium-1-olate
Molecular Formula
C5H3BrN2O3
InChI Key
YZQMKADIENBVLH-UHFFFAOYSA-N
SMILES
[O-][N+](=O)C1=CC=[N+]([O-])C=C1Br
Appearance (Color)
Yellow
Form
Crystals or powder or crystalline powder
Melting Point (clear melt)
151.0-157.0?C
Assay (GC)
≥98.0%

Description

3-Bromo-4-nitropyridine N-oxide is an important raw material and intermediate used in organic synthesis, pharmaceuticals agrochemicals and dyestuff fields.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Bromo-4-nitropyridine N-oxide is an important raw material and intermediate used in organic synthesis, pharmaceuticals agrochemicals and dyestuff fields.

Solubility
Sparingly Soluble in water (0.92 g/L at 25°C).

Notes
Store under inert gas. It is sensitive to air. Store in a cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text