Thermo Scientific Chemicals

2,3-Dibromopyridine, 97%, Thermo Scientific Chemicals

Catalog number: L19527.14
25 g, Each
Thermo Scientific Chemicals

2,3-Dibromopyridine, 97%, Thermo Scientific Chemicals

Catalog number: L19527.14
25 g, Each
Quantity
Catalog number: L19527.14
also known as L19527-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
13534-89-9
IUPAC Name
2,3-dibromopyridine
Molecular Formula
C5H3Br2N
InChI Key
SLMHHOVQRSSRCV-UHFFFAOYSA-N
SMILES
BrC1=CC=CN=C1Br
Form
Powder
Appearance (Color)
White to cream to pale brown
Assay (GC)
≥96.0%
Melting Point (clear melt)
55.0-64.0?C

Description

It is employed as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed as a pharmaceutical intermediate.

Solubility
Insoluble in water.

Notes
Moisture sensitive. Store in dry condition. Keep oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text