3-Cyano-2-fluoropyridine, 98%, Thermo Scientific Chemicals
3-Cyano-2-fluoropyridine, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

3-Cyano-2-fluoropyridine, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number L19538.03
also known as L19538-03
Price (USD)/ Each
29.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
34.60 
Save 4.95 (14%)
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)/ Each
29.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
34.60 
Save 4.95 (14%)
Add to cart
3-Cyano-2-fluoropyridine, 98%, Thermo Scientific Chemicals
Catalog numberL19538.03
Price (USD)/ Each
29.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
34.60 
Save 4.95 (14%)
-
Add to cart
Chemical Identifiers
CAS3939-13-7
IUPAC Name2-fluoropyridine-3-carbonitrile
Molecular FormulaC6H3FN2
InChI KeyUSIDQCCXMGJOJM-UHFFFAOYSA-N
SMILESFC1=NC=CC=C1C#N
View more
SpecificationsSpecification SheetSpecification Sheet
Melting Point (clear melt)25.0-31.0?C
Appearance (Color)Colorless to white
Assay (GC)≥97.5%
FormCrystals or powder or crystalline powder or clear liquid as melt
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only