2,3,6-Trifluoropyridine, 97%, Thermo Scientific Chemicals
2,3,6-Trifluoropyridine, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2,3,6-Trifluoropyridine, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Catalog number L19540.06
also known as L19540-06
Price (USD)/ Each
126.65
Online exclusive
141.00 
Save 14.35 (10%)
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
126.65
Online exclusive
141.00 
Save 14.35 (10%)
Add to cart
2,3,6-Trifluoropyridine, 97%, Thermo Scientific Chemicals
Catalog numberL19540.06
Price (USD)/ Each
126.65
Online exclusive
141.00 
Save 14.35 (10%)
-
Add to cart
Chemical Identifiers
CAS3512-18-3
IUPAC Name2,3,6-trifluoropyridine
Molecular FormulaC5H2F3N
InChI KeyHRLIANGJWPJFKW-UHFFFAOYSA-N
SMILESFC1=CC=C(F)C(F)=N1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow to orange to red
Refractive Index1.4180-1.4230 @ 20?C
FormLiquid
Identification (FTIR)Conforms
Assay (GC)≥96.0%
2,3,6-Trifluoropyridine is used as an intermediate in organic syntheses and in pharmaceuticals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,3,6-Trifluoropyridine is used as an intermediate in organic syntheses and in pharmaceuticals.

Solubility
Not miscible or difficult to mix in water.

Notes
Keep the container tightly sealed. Store in a cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
RUO – Research Use Only

General References:

  1. Sergey S Laeva, Vladimir U Evtefeevb, Vitalij D Shteingarts. A new approach to polyfluoroaromatic amines with an unsubstituted position ortho to the amino group. Journal of Fluorine Chemistry. 2001, 110 (1), 43-46.
  2. Yoon-Joo Ko, Kyung-Bae Park, Seung-Bo Shim, Jung-Hyu Shin. Synthesis of difluorinated pyridinecarboxaldehyde via electrophilic fluorination. Journal of Fluorine Chemistry. 2006, 127 (6), 755-759.