2,4,6-Trifluoropyridine, 97%, Thermo Scientific Chemicals
2,4,6-Trifluoropyridine, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2,4,6-Trifluoropyridine, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number L19541.06
also known as L19541-06
Price (USD)/ Each
130.65
Online exclusive
145.00 
Save 14.35 (10%)
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
130.65
Online exclusive
145.00 
Save 14.35 (10%)
Add to cart
2,4,6-Trifluoropyridine, 97%, Thermo Scientific Chemicals
Catalog numberL19541.06
Price (USD)/ Each
130.65
Online exclusive
145.00 
Save 14.35 (10%)
-
Add to cart
Chemical Identifiers
CAS3512-17-2
IUPAC Name2,4,6-trifluoropyridine
Molecular FormulaC5H2F3N
InChI KeyUZDRWXKBKVVUTE-UHFFFAOYSA-N
SMILESFC1=CC(F)=NC(F)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Assay (GC)≥96.0%
Identification (FTIR)Conforms
Refractive Index1.4100-1.4150 @ 20?C
Appearance (Color)Clear colorless
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only