Thermo Scientific Chemicals

2-(Trifluoromethyl)pyridine, 99%, Thermo Scientific Chemicals

Catalog number: L19554.03
1 g, Each
Thermo Scientific Chemicals

2-(Trifluoromethyl)pyridine, 99%, Thermo Scientific Chemicals

Catalog number: L19554.03
1 g, Each
Quantity
Catalog number: L19554.03
also known as L19554-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
368-48-9
Refractive Index
1.4170-1.4200 @ 20?C
Assay (GC)
≥98.5%
Appearance (Color)
Clear colorless to pale yellow
Form
Liquid

Description

Lithiation with n-BuLi/LiOCH2CH2NMe2 takes place at the 6-position, affording access to 6-substituted derivatives. With LiTMP . It is a building block in heterocycle synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Lithiation with n-BuLi/LiOCH2CH2NMe2 takes place at the 6-position, affording access to 6-substituted derivatives. With LiTMP . It is a building block in heterocycle synthesis.

Solubility
Soluble in most organic solvents.

Notes
Store at room temperature. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text