Thermo Scientific Chemicals

Amberlite XAD-7, Thermo Scientific Chemicals

Catalog number: L19564.22
100 g, Each
Thermo Scientific Chemicals

Amberlite XAD-7, Thermo Scientific Chemicals

Catalog number: L19564.22
100 g, Each
Quantity
Catalog number: L19564.22
also known as L19564-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
37380-43-1
MDL Number
MFCD00132705
Particle Size
Uniformity Coefficient: ≤ 2.0
Appearance (Color)
White to pink-beige
Comment
Product can absorb non-polar compounds from aqueous systems, and can absorb polar compounds from non-polar solvents.
Harmonic mean size
0.430-0.690mm
Form
Beads

Description

Amberlite XAD-7 is used in chromatography media, enzyme immobilization and water purification. It is also used in the removal of relatively polar compounds from non aqueous solvents, non-aromatic compounds from polar solvent, recovery of plant extracts, antibiotics, enzymes and proteins.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Amberlite XAD-7 is used in chromatography media, enzyme immobilization and water purification. It is also used in the removal of relatively polar compounds from non aqueous solvents, non-aromatic compounds from polar solvent, recovery of plant extracts, antibiotics, enzymes and proteins.

Solubility
Insoluble in water.

Notes
Keep the container tightly sealed. Store in a cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text