Thermo Scientific Chemicals

Amberlite XAD-16, Thermo Scientific Chemicals

Catalog number: L19565.22
100 g, Each
Thermo Scientific Chemicals

Amberlite XAD-16, Thermo Scientific Chemicals

Catalog number: L19565.22
100 g, Each
Quantity
Catalog number: L19565.22
also known as L19565-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
104219-63-8
Form
Beads
Residual water
62 - 70%
Particle Size
≤ 2.0% @ < 0.300mm
Particle Size
Harmonic mean 0.560 - 0.710mm
Appearance (Color)
White to cream

Description

Amberlite XAD-16 is a polymeric adsorbent widely utilized for adsorption of organic substances from aqueous systems and polar solvents. It is useful for the removal of detergents from protein solutions. It is also used in the recovery and purification of antibiotics, water soluble steroids, amino acids and proteins. It plays a vital role in the removal of non polar compounds from polar solvents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Amberlite XAD-16 is a polymeric adsorbent widely utilized for adsorption of organic substances from aqueous systems and polar solvents. It is useful for the removal of detergents from protein solutions. It is also used in the recovery and purification of antibiotics, water soluble steroids, amino acids and proteins. It plays a vital role in the removal of non polar compounds from polar solvents.

Solubility
Insoluble in water.

Notes
Incompatible with strong oxidizing agents and acids.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text