Thermo Scientific Chemicals

Amberlite™ IRC-748, ion exchange resin, Thermo Scientific Chemicals

Catalog number: L19570.30
250 g, Each
Thermo Scientific Chemicals

Amberlite™ IRC-748, ion exchange resin, Thermo Scientific Chemicals

Catalog number: L19570.30
250 g, Each
Quantity
Catalog number: L19570.30
also known as L19570-30
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
79620-28-3
MDL Number
MFCD00132702
Comment
Moisture holding capacity: 60-65% (Na form)
Form
Beads or pellets
Appearance (Color)
Cream to pale brown
Comment
Exchange capacity: ≥1.35eq/L (Na form)
Particle Size
0.50-0.65mm

Description

Amberlite™ IRC-748, ion exchange resin is used as a chelating resin with high affinity for heavy metal cations compared to alkali or alkaline earth metals. It is also utilized in metal-chelation applications. Further, it is used in purification of brine in the chloralkali industry as well as the purification of process streams having trace metals. In addition to this, it is used in the electroplating industry and in the recovery of heavy metals from hydrometallurgical leach streams.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Amberlite|r IRC-748, ion exchange resin is used as a chelating resin with high affinity for heavy metal cations compared to alkali or alkaline earth metals. It is also utilized in metal-chelation applications. Further, it is used in purification of brine in the chloralkali industry as well as the purification of process streams having trace metals. In addition to this, it is used in the electroplating industry and in the recovery of heavy metals from hydrometallurgical leach streams.

Solubility
Insoluble in water.

Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text