Thermo Scientific Chemicals

(R)-(-)-2-Chloromandelic acid, ChiPros 99+%, ee 99+%, Thermo Scientific Chemicals

Catalog number: L19586.06
5 g, Each
Thermo Scientific Chemicals

(R)-(-)-2-Chloromandelic acid, ChiPros 99+%, ee 99+%, Thermo Scientific Chemicals

Catalog number: L19586.06
5 g, Each
Quantity
Catalog number: L19586.06
also known as L19586-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
52950-18-2
IUPAC Name
2-(2-chlorophenyl)-2-hydroxyacetic acid
Molecular Formula
C8H7ClO3
InChI Key
RWOLDZZTBNYTMS-UHFFFAOYNA-N
SMILES
OC(C(O)=O)C1=CC=CC=C1Cl
Appearance (Color)
White
Form
crystalline powder
Assay (Aqueous acid-base Titration)
>99.0%
Enantiomeric excess
>99.0%
Optical Rotation
-123 ? 2? (c=3 in water)

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text