Thermo Scientific Chemicals

2-Aminopyridine-3-carboxaldehyde, 98%, Thermo Scientific Chemicals

Catalog number: L19620.03
1 g, Each
Thermo Scientific Chemicals

2-Aminopyridine-3-carboxaldehyde, 98%, Thermo Scientific Chemicals

Catalog number: L19620.03
1 g, Each
Quantity
Catalog number: L19620.03
also known as L19620-03
Price (EUR)
Quantity
-

Chemical Identifiers

CAS
7521-41-7
IUPAC Name
2-amino-3-formylpyridin-1-ium
Molecular Formula
C6H7N2O
InChI Key
NXMFJCRMSDRXLD-UHFFFAOYSA-O
SMILES
NC1=[NH+]C=CC=C1C=O
Appearance (Color)
Yellow
Form
Crystalline powder
Assay (GC)
≥97.5%
Free acid (titration)
<1.0%
Identification (FTIR)
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text

    Safety and Handling


    Classification of the substance or mixture
    CLP classification - Regulation(EC) No 1272/2008
    Skin Corrosion/Irritation
    Category 2
    Serious Eye Damage/Eye Irritation
    Category 2
    Specific target organ toxicity - (single exposure)
    Category 3
    Label Elements
    Signal Word

    Warning

    Hazard Statements

    H315 - Causes skin irritation

    H319 - Causes serious eye irritation

    H335 - May cause respiratory irritation

    Precautionary Statements

    P280 - Wear protective gloves/protective clothing/eye protection/face protection

    P305 + P351 + P338 - IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing