3-Bromo-4-methylpyridine, 98%, Thermo Scientific Chemicals
3-Bromo-4-methylpyridine, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

3-Bromo-4-methylpyridine, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Catalog number L19622.14
also known as L19622-14
Price (USD)/ Each
397.00
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
397.00
Add to cart
3-Bromo-4-methylpyridine, 98%, Thermo Scientific Chemicals
Catalog numberL19622.14
Price (USD)/ Each
397.00
-
Add to cart
Chemical Identifiers
CAS3430-22-6
IUPAC Name3-bromo-4-methylpyridine
Molecular FormulaC6H6BrN
InChI KeyGSQZOLXWFQQJHJ-UHFFFAOYSA-N
SMILESCC1=CC=NC=C1Br
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow
FormLiquid
Assay (GC)≥97.5%
Refractive Index1.5605-1.5645 @ 20?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only