4-Methoxypyridine, 98+%, Thermo Scientific Chemicals
4-Methoxypyridine, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-Methoxypyridine, 98+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Catalog number L19625.14
also known as L19625-14
Price (USD)/ Each
250.00
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
250.00
Add to cart
4-Methoxypyridine, 98+%, Thermo Scientific Chemicals
Catalog numberL19625.14
Price (USD)/ Each
250.00
-
Add to cart
Chemical Identifiers
CAS620-08-6
IUPAC Name4-methoxypyridine
Molecular FormulaC6H7NO
InChI KeyXQABVLBGNWBWIV-UHFFFAOYSA-N
SMILESCOC1=CC=NC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to pale yellow
FormLiquid
Assay (GC)≥98.0%
CommentMay darken on storage
Refractive Index1.5165-1.5235 @ 20?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only