4-(Methylamino)pyridine, 99%, Thermo Scientific Chemicals
4-(Methylamino)pyridine, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-(Methylamino)pyridine, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number L19626.03
also known as L19626-03
Price (USD)/ Each
46.10
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)/ Each
46.10
Add to cart
4-(Methylamino)pyridine, 99%, Thermo Scientific Chemicals
Catalog numberL19626.03
Price (USD)/ Each
46.10
-
Add to cart
Chemical Identifiers
CAS1121-58-0
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder
Assay (GC)≥98.5%
Melting Point (clear melt)123.0-129.0?C
Appearance (Color)White to cream
4-(Methylamino)pyridine is employed as efficient nucleophilic catalyst during the preparation of ultra-high surface area emulsion templated porous polymers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-(Methylamino)pyridine is employed as efficient nucleophilic catalyst during the preparation of ultra-high surface area emulsion templated porous polymers.

Solubility
Soluble in water, acetone, ethanol, ether and benzene.

Notes
Keep the container tightly sealed. Store in a cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
RUO – Research Use Only

General References:

  1. Kin-Shan Huang.; Doyle Britton.; Stephen R. Byrn. A novel class of phenol-pyridine co-crystals for secondharmonic generation. Journal of Materials Chemistry. 1997, 7 (5), 713-720.
  2. Irena Pulko Dr.; Jennifer Wall.; Peter Krajnc Prof.; Neil R. Cameron Prof. Ultra-High Surface Area Functional Porous Polymers by Emulsion Templating and Hypercrosslinking: Efficient Nucleophilic Catalyst Supports. Chemistry - A European Journal. 2010, 16 (8), 2350-2354.