Thermo Scientific Chemicals

Isoquinoline-5-carboxylic acid, 96%, Thermo Scientific Chemicals

Catalog number: L19637.MD
250 mg, Each
Thermo Scientific Chemicals

Isoquinoline-5-carboxylic acid, 96%, Thermo Scientific Chemicals

Catalog number: L19637.MD
250 mg, Each
Quantity
Catalog number: L19637.MD
also known as L19637-MD
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
27810-64-6
IUPAC Name
isoquinoline-5-carboxylic acid
Molecular Formula
C10H7NO2
InChI Key
ZIPLFLRGHZAXSJ-UHFFFAOYSA-N
SMILES
OC(=O)C1=C2C=CN=CC2=CC=C1
Appearance (Color)
White to cream to pale brown to yellow
Assay (Non-aqueous acid-base Titration)
≥95.0%
Form
Powder

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text