Quinoline-8-carboxaldehyde is an important raw material and intermediate used in organic synthesis, pharmaceuticals agrochemicals and dyestuff fields.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Quinoline-8-carboxaldehyde is an important raw material and intermediate used in organic synthesis, pharmaceuticals agrochemicals and dyestuff fields.
Solubility
Insoluble in water.
Notes
Keep the container tightly sealed. Store in a cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
RUO – Research Use Only
General References:
- Clemens G. Anklin.; Paul S. Pregosin.; Franz J. Wombacher.; Hubert J. Ruegg. Mechanistic aspects of the cyclometalation of quinoline-8-carboxaldehyde with platinum(II). Organometallics. 1990, 9 (6), 1953-1958.
- Katarzyna Guzow.; Magda Milewska.; Dominik Wróblewski.; Artur Giełdoń.; Wiesław Wiczk. 3-[2-(8-Quinolinyl)benzoxazol-5-yl]alanine derivative—a specific fluorophore for transition and rare-earth metal ion detection. Tetrahedron. 2004, 60 (51), 11889-11894.