Thermo Scientific Chemicals

Quinoline-5-carboxylic acid, 97+%, Thermo Scientific Chemicals

Catalog number: L19643.MD
250 mg, Each
Thermo Scientific Chemicals

Quinoline-5-carboxylic acid, 97+%, Thermo Scientific Chemicals

Catalog number: L19643.MD
250 mg, Each
Quantity
Catalog number: L19643.MD
also known as L19643-MD
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7250-53-5
Appearance (Color)
White to cream to pale brown to pale gray
Form
Powder
Assay (Non-aqueous acid-base Titration)
≥97.0 to ≤103.0%
Assay (HPLC)
≥97.0%
Melting Point (clear melt)
295.0-305.0?C

Description

Quinoline-5-carboxylic acid is an important raw material and intermediate used in organic synthesis, pharmaceuticals agrochemicals and dyestuff fields.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Quinoline-5-carboxylic acid is an important raw material and intermediate used in organic synthesis, pharmaceuticals agrochemicals and dyestuff fields.

Solubility
Slightly soluble in water.

Notes
Keep the container tightly sealed. Store in a cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text