Thermo Scientific Chemicals

1H-Pyrazole-4-boronic acid pinacol ester, 98%, Thermo Scientific Chemicals

Catalog number: L19654.06
5 g, Each
Thermo Scientific Chemicals

1H-Pyrazole-4-boronic acid pinacol ester, 98%, Thermo Scientific Chemicals

Catalog number: L19654.06
5 g, Each
Quantity
Catalog number: L19654.06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
269410-08-4
IUPAC Name
4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole
Molecular Formula
C9H15BN2O2
InChI Key
TVOJIBGZFYMWDT-UHFFFAOYSA-N
SMILES
CC1(C)OB(OC1(C)C)C1=CNN=C1
Form
Crystals or powder or crystalline powder
Assay (GC)
≥97.5%
Appearance (Color)
White to cream to pale brown

Description

1H-Pyrazole-4-boronic acid pinacol ester is a useful reagent for Suzuki-Miyaura cross-couplings as well as Ruthenium-catalyzed asymmetric hydrogenation. It is also used as reagent for preparing VEGF, Aurora, RHO (ROCK), Janus Kinase 2, c-MET, ALK, S-nitrsoflutathione reductase, CDC7, Acetyl-CoA carboxylase inhibitors as well as other biologically active compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1H-Pyrazole-4-boronic acid pinacol ester is a useful reagent for Suzuki-Miyaura cross-couplings as well as Ruthenium-catalyzed asymmetric hydrogenation. It is also used as reagent for preparing VEGF, Aurora, RHO (ROCK), Janus Kinase 2, c-MET, ALK, S-nitrsoflutathione reductase, CDC7, Acetyl-CoA carboxylase inhibitors as well as other biologically active compounds.

Solubility
Insoluble in water.

Notes
Keep the container tightly sealed. Store in a cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text