5-Iodo-2-furaldehyde, 97%, Thermo Scientific Chemicals
5-Iodo-2-furaldehyde, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

5-Iodo-2-furaldehyde, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Catalog number L19670.06
also known as L19670-06
Price (USD)/ Each
165.00
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
165.00
Add to cart
5-Iodo-2-furaldehyde, 97%, Thermo Scientific Chemicals
Catalog numberL19670.06
Price (USD)/ Each
165.00
-
Add to cart
Chemical Identifiers
CAS2689-65-8
IUPAC Name5-iodofuran-2-carbaldehyde
Molecular FormulaC5H3IO2
InChI KeyQPGPCPKDVSPJAY-UHFFFAOYSA-N
SMILESIC1=CC=C(O1)C=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥96.0%
Free acid (titration)≤2.0%
Melting Point (clear melt)123.5-129.5?C
FormCrystals or powder or crystalline powder
Appearance (Color)Orange
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only