4-Fluoro-3-methoxybenzaldehyde, 98%, Thermo Scientific Chemicals
4-Fluoro-3-methoxybenzaldehyde, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-Fluoro-3-methoxybenzaldehyde, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number L19671.03
also known as L19671-03
Price (USD)/ Each
42.40
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)/ Each
42.40
Add to cart
4-Fluoro-3-methoxybenzaldehyde, 98%, Thermo Scientific Chemicals
Catalog numberL19671.03
Price (USD)/ Each
42.40
-
Add to cart
Chemical Identifiers
CAS128495-46-5
IUPAC Name4-fluoro-3-methoxybenzaldehyde
Molecular FormulaC8H7FO2
InChI KeyNALVGTOMKSKFFV-UHFFFAOYSA-N
SMILESCOC1=CC(C=O)=CC=C1F
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale cream to cream to yellow
FormCrystals or powder or crystalline powder or fused solid
Assay (GC)≥97.5%
Free acid (titration)≤1%
Melting Point (clear melt)57.0-64.0?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only