Thermo Scientific Chemicals

(R)-(-)-1-Benzyl-3-aminopyrrolidine, 99%, ee 99%, Thermo Scientific Chemicals

Catalog number: L19690.06
5 g, Each
Thermo Scientific Chemicals

(R)-(-)-1-Benzyl-3-aminopyrrolidine, 99%, ee 99%, Thermo Scientific Chemicals

Catalog number: L19690.06
5 g, Each
Quantity
Catalog number: L19690.06
also known as L19690-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
114715-39-8
IUPAC Name
(3R)-1-benzylpyrrolidin-3-amine
Molecular Formula
C11H16N2
InChI Key
HBVNLKQGRZPGRP-LLVKDONJSA-N
SMILES
N[C@@H]1CCN(CC2=CC=CC=C2)C1
Assay (GC)
≥98.5%
Appearance (Color)
Clear colourless to yellow
Comment
Optical rotation is ca. -2° (neat) (non-U.S. specification)
Form
Liquid
Refractive Index
1.5410-1.5450 @20°C (non-U.S. specification)

Description

(R)-(-)-1-Benzyl-3-aminopyrrolidine is a important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(R)-(-)-1-Benzyl-3-aminopyrrolidine is a important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field.

Solubility
Slightly soluble in water.

Notes
Air sensitive. Store under inert gas. Store away from oxidizing agents, air, carbondioxide.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text