3-Acetoxy-1-propenylboronic acid pinacol ester, 97%, Thermo Scientific Chemicals
3-Acetoxy-1-propenylboronic acid pinacol ester, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

3-Acetoxy-1-propenylboronic acid pinacol ester, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Catalog number L19700.MD
also known as L19700-MD
Price (USD)/ Each
119.00
-
Add to cart
Quantity:
250 mg
Request bulk or custom format
Price (USD)/ Each
119.00
Add to cart
3-Acetoxy-1-propenylboronic acid pinacol ester, 97%, Thermo Scientific Chemicals
Catalog numberL19700.MD
Price (USD)/ Each
119.00
-
Add to cart
Chemical Identifiers
CAS161395-97-7
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to pale yellow
FormLiquid
Assay (GC)≥96.0%
Refractive Index1.4450-1.4500 @ 20?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only