Thermo Scientific Chemicals

2-Bromo-4-fluorobenzenesulfonyl chloride, 97%, Thermo Scientific Chemicals

Catalog number: L19713.03
1 g, Each
Thermo Scientific Chemicals

2-Bromo-4-fluorobenzenesulfonyl chloride, 97%, Thermo Scientific Chemicals

Catalog number: L19713.03
1 g, Each
Quantity
Catalog number: L19713.03
also known as L19713-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
351003-45-7
IUPAC Name
2-bromo-4-fluorobenzene-1-sulfonyl chloride
Molecular Formula
C6H3BrClFO2S
InChI Key
GJPWPFVLPNTOOL-UHFFFAOYSA-N
SMILES
FC1=CC(Br)=C(C=C1)S(Cl)(=O)=O
Appearance (Color)
Colorless to pale cream to pale yellow
Refractive Index
1.5750-1.5800 @ 20?C
Assay (GC)
≥96.0%
Melting Point (clear melt)
33.0-42.0?C
Form
Fused solid or clear liquid as melt

Description

2-Bromo-4-fluorobenzenesulfonyl chloride is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Bromo-4-fluorobenzenesulfonyl chloride is used as a pharmaceutical intermediate.

Solubility
Hydrolyzes in water.

Notes
Moisture sensitive. Store away from oxidizing agents, water/ moisture, bases.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text