Thermo Scientific Chemicals

4-Cyano-2-fluorobenzoic acid, 98%, Thermo Scientific Chemicals

Catalog number: L19722.14
25 g, Each
Thermo Scientific Chemicals

4-Cyano-2-fluorobenzoic acid, 98%, Thermo Scientific Chemicals

Catalog number: L19722.14
25 g, Each
Quantity
Catalog number: L19722.14
also known as L19722-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
164149-28-4
IUPAC Name
4-cyano-2-fluorobenzoic acid
Molecular Formula
C8H4FNO2
InChI Key
KEJMSTJTAWACNI-UHFFFAOYSA-N
SMILES
OC(=O)C1=C(F)C=C(C=C1)C#N
Melting Point (clear melt)
214.0-220.0?C
Form
Crystals or powder or crystalline powder
Assay (Silylated GC)
≥97.5%
Appearance (Color)
White
Assay (Aqueous acid-base Titration)
≥97.5 to ≤102.5%

Description

4-Cyano-2-fluorobenzoic acid is an important raw material and intermediate used in organic synthesis, pharmaceuticals agrochemicals and dyestuff fields.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Cyano-2-fluorobenzoic acid is an important raw material and intermediate used in organic synthesis, pharmaceuticals agrochemicals and dyestuff fields.

Solubility
Insoluble in water.

Notes
Keep the container tightly sealed. Store in a cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text