4-Amino-3-nitropyridine, 96%, Thermo Scientific Chemicals
4-Amino-3-nitropyridine, 96%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-Amino-3-nitropyridine, 96%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number L19725.06
also known as L19725-06
Price (USD)/ Each
101.00
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
101.00
Add to cart
4-Amino-3-nitropyridine, 96%, Thermo Scientific Chemicals
Catalog numberL19725.06
Price (USD)/ Each
101.00
-
Add to cart
Chemical Identifiers
CAS1681-37-4
IUPAC Name4-amino-3-nitropyridin-1-ium
Molecular FormulaC5H6N3O2
InChI KeyIUPPEELMBOPLDJ-UHFFFAOYSA-O
SMILESNC1=CC=[NH+]C=C1[N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Dark yellow to yellow
Assay (Non-aqueous acid-base Titration)≥95.0 to ≤105.0%
FormPowder
Melting Point (clear melt)198.0-208.0?C
4-Amino-3-nitropyridine is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Amino-3-nitropyridine is used as a pharmaceutical intermediate.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agents.
RUO – Research Use Only

General References:

  1. J. W. Clark-Lewis and R. P. Singh. 459. Preparation of 3,4-diamino-, 3-amino-4-methylamino-, and 4-amino-3-methylamino-pyridine. J. Med. Chem. 1962, 2379-2382.
  2. Samir Sabbah, Gerhard K.E Scriba. Development and validation of a capillary electrophoresis assay for the determination of 3,4-diaminopyridine and 4-aminopyridine including related substances. Journal of Chromatography A. 2001, 907, (1-2), 321-328.