Methyl 3,3-dimethylacrylate is an important raw material and intermediate used in organic synthesis, pharmaceuticals agrochemicals and dyestuff fields.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Methyl 3,3-dimethylacrylate is an important raw material and intermediate used in organic synthesis, pharmaceuticals agrochemicals and dyestuff fields.
Solubility
Soluble in chloroform. Insoluble in water,
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents. It is sensitive to light.
RUO – Research Use Only
Yasutsugu Ueda.; Amarendra B. Mikkilineni.; Jay O. Knipe.; William C. Rose.; Anna Maria Casazza.; Dolatrai M. Vyas. Novel water soluble phosphate prodrugs of taxol™ possessing in vivo antitumor activity. Bioorganic & Medicinal Chemistry Letters. 1993, 3 (8), 1761-1766.
Jack E. Baldwin.; Stephen B. Haber, Carolyn Hoskins, Lawrence I. Kruse. Synthesis of .beta.,.gamma.-unsaturated amino acids. J. Org. Chem. 1977, 42 (7), 1239-1241.