Thermo Scientific Chemicals

1-n-Butyl-3-methylimidazolium chloride, 96%, Thermo Scientific Chemicals

Catalog number: L19749.09
10 g, Each
Thermo Scientific Chemicals

1-n-Butyl-3-methylimidazolium chloride, 96%, Thermo Scientific Chemicals

Catalog number: L19749.09
10 g, Each
Quantity
Catalog number: L19749.09
also known as L19749-09
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
79917-90-1
IUPAC Name
1-butyl-3-methyl-1H-imidazol-3-ium chloride
Molecular Formula
C8H15ClN2
InChI Key
FHDQNOXQSTVAIC-UHFFFAOYSA-M
SMILES
[Cl-].CCCCN1C=C[N+](C)=C1
Appearance (Color)
White to cream to pale yellow or pale brown
Assay (Titration ex Chloride)
≥95.0 to ≤105.0%
Identification (FTIR)
Conforms
Form
Crystals or powder or crystalline powder or lumps
Assay (HPLC)
≥95.0%

Description

1-n-Butyl-3-methylimidazolium chloride is used as a solvent for organic reactions. It is used in anion exchange reactions. It acts as a catalyst, electrolyte for batteries and metal recovery. It is also used in chromatographic separations. Further, it is used as an ionic liquids for catalysis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-n-Butyl-3-methylimidazolium chloride is used as a solvent for organic reactions. It is used in anion exchange reactions. It acts as a catalyst, electrolyte for batteries and metal recovery. It is also used in chromatographic separations. Further, it is used as an ionic liquids for catalysis.

Solubility
Soluble in acetone, acetonitrile, hot ethyl acetate, isopropyl alcohol, methylene chloride and methanol. Insoluble in water, hexane and toluene,

Notes
Hygroscopic. Moisture sensitive. Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text