Thermo Scientific Chemicals

4-Morpholinylsulfur trifluoride, 95%, Thermo Scientific Chemicals

Catalog number: L19751.06
5 g, Each
Thermo Scientific Chemicals

4-Morpholinylsulfur trifluoride, 95%, Thermo Scientific Chemicals

Catalog number: L19751.06
5 g, Each
Quantity
Catalog number: L19751.06
also known as L19751-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
51010-74-3
IUPAC Name
4-(trifluoro-λ⁴-sulfanyl)morpholine
Molecular Formula
C4H8F3NOS
InChI Key
UFXIRMVZNARBDL-UHFFFAOYSA-N
SMILES
FS(F)(F)N1CCOCC1
Appearance (Color)
Clear colorless to yellow
Refractive Index
1.4515-1.4575 @ 20?C
Assay (Non-aqueous acid-base Titration)
≥94.0 to ≤106.0%
Form
Liquid

Description

4-Morpholinylsulfur trifluoride is incorporated in the fluoronation of trihexanols. It is used as a nucleophilic fluorinating agent with greater thermal stability. It is also used in the preparation of proline analogs and in the preparation of difluoropiperidines.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Morpholinylsulfur trifluoride is incorporated in the fluoronation of trihexanols. It is used as a nucleophilic fluorinating agent with greater thermal stability. It is also used in the preparation of proline analogs and in the preparation of difluoropiperidines.

Solubility
Reacts with water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with bases and oxidizing and bases. Store under dry inert gas. It is sensitive to moisture.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text