1-Ethyl-3-methylimidazolium hexafluorophosphate, 98+%, Thermo Scientific Chemicals
1-Ethyl-3-methylimidazolium hexafluorophosphate, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

1-Ethyl-3-methylimidazolium hexafluorophosphate, 98+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
2 g
10 g
50 g
Catalog number L19762.04
also known as L19762-04
Price (USD)/ Each
61.70
-
Add to cart
Quantity:
2 g
Request bulk or custom format
Price (USD)/ Each
61.70
Add to cart
1-Ethyl-3-methylimidazolium hexafluorophosphate, 98+%, Thermo Scientific Chemicals
Catalog numberL19762.04
Price (USD)/ Each
61.70
-
Add to cart
Chemical Identifiers
CAS155371-19-0
IUPAC Name1-ethyl-3-methyl-1H-imidazol-3-ium; hexafluoro-λ⁵-phosphanuide
Molecular FormulaC6H11F6N2P
InChI KeyDPDAKOVGQUGTHH-UHFFFAOYSA-N
SMILESF[P-](F)(F)(F)(F)F.CCN1C=C[N+](C)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White or colorless to yellow
Assay from Supplier's CofA≥98.0% (H-NMR)
FormCrystalline or fused solid
Water Content (Karl Fischer Titration)< 1.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only