Thermo Scientific Chemicals

1-Ethyl-3-methylimidazolium hexafluorophosphate, 98+%, Thermo Scientific Chemicals

Catalog number: L19762.18
50 g, Each
Thermo Scientific Chemicals

1-Ethyl-3-methylimidazolium hexafluorophosphate, 98+%, Thermo Scientific Chemicals

Catalog number: L19762.18
50 g, Each
Quantity
Catalog number: L19762.18
also known as L19762-18
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
155371-19-0
IUPAC Name
1-ethyl-3-methyl-1H-imidazol-3-ium; hexafluoro-λ⁵-phosphanuide
Molecular Formula
C6H11F6N2P
InChI Key
DPDAKOVGQUGTHH-UHFFFAOYSA-N
SMILES
F[P-](F)(F)(F)(F)F.CCN1C=C[N+](C)=C1
Appearance (Color)
White or colorless to yellow
Assay from Supplier's CofA
≥98.0% (H-NMR)
Form
Crystalline or fused solid
Water Content (Karl Fischer Titration)
< 1.0%

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text