Thermo Scientific Chemicals

1-Ethyl-3-methylimidazolium tetrafluoroborate, 98+% (dry wt.), may cont. up to 3% water, Thermo Scientific Chemicals

Catalog number: L19763.09
10 g, Each
Thermo Scientific Chemicals

1-Ethyl-3-methylimidazolium tetrafluoroborate, 98+% (dry wt.), may cont. up to 3% water, Thermo Scientific Chemicals

Catalog number: L19763.09
10 g, Each
Quantity
Catalog number: L19763.09
also known as L19763-09
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
143314-16-3
IUPAC Name
1-ethyl-3-methyl-1H-imidazol-3-ium; tetrafluoroboranuide
Molecular Formula
C6H11BF4N2
InChI Key
CUNYTRQQXKCRTJ-UHFFFAOYSA-N
SMILES
F[B-](F)(F)F.CCN1C=C[N+](C)=C1
Appearance (Color)
Clear colorless to orange
Assay from Supplier's CofA
≥98.0% (NMR)
Assay from Supplier's CofA
Halides: ≤1% (IC)
Water Content (Karl Fischer Titration)
<3.0%
Assay from Supplier's CofA
1-Ethyl-3-methylimidazolium: ≥98.0% (IC)

Description

1-Ethyl-3-methylimidazolium tetrafluoroborate is an ionic liquid used in the recycling of osmium in the dihydroxylation of olefins with osmium(VIII)¬ oxide. It is a stable liquid electrolyte used in the capillary electrophoretic method for resolving phenolic compounds present in grape seed extract. It also acts as a good solvent for a wide range of both organic and inorganic compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Ethyl-3-methylimidazolium tetrafluoroborate is an ionic liquid used in the recycling of osmium in the dihydroxylation of olefins with osmium(VIII)¬ oxide. It is a stable liquid electrolyte used in the capillary electrophoretic method for resolving phenolic compounds present in grape seed extract. It also acts as a good solvent for a wide range of both organic and inorganic compounds.

Notes
Hygroscopic. Incompatible with strong oxidizing agents and water.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text