3-Cyano-4-methylpyridine, 97%, Thermo Scientific Chemicals
3-Cyano-4-methylpyridine, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

3-Cyano-4-methylpyridine, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Catalog number L19769.06
also known as L19769-06
Price (USD)/ Each
116.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
137.00 
Save 20.35 (15%)
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
116.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
137.00 
Save 20.35 (15%)
Add to cart
3-Cyano-4-methylpyridine, 97%, Thermo Scientific Chemicals
Catalog numberL19769.06
Price (USD)/ Each
116.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
137.00 
Save 20.35 (15%)
-
Add to cart
Chemical Identifiers
CAS5444-01-9
IUPAC Name4-methylpyridine-3-carbonitrile
Molecular FormulaC7H6N2
InChI KeyXLAPHZHNODDMDD-UHFFFAOYSA-N
SMILESCC1=CC=NC=C1C#N
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder or lumps
Melting Point (clear melt)38.0-47.0?C
Appearance (Color)White to cream to yellow to brown
Assay (GC)≥96.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only