Ethyl (R)-(-)-4-cyano-3-hydroxybutyrate, 98%, Thermo Scientific Chemicals
Ethyl (R)-(-)-4-cyano-3-hydroxybutyrate, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Ethyl (R)-(-)-4-cyano-3-hydroxybutyrate, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Catalog number L19771.14
also known as L19771-14
Price (USD)/ Each
161.00
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
161.00
Add to cart
Ethyl (R)-(-)-4-cyano-3-hydroxybutyrate, 98%, Thermo Scientific Chemicals
Catalog numberL19771.14
Price (USD)/ Each
161.00
-
Add to cart
Chemical Identifiers
CAS141942-85-0
IUPAC Nameethyl (3R)-4-cyano-3-hydroxybutanoate
Molecular FormulaC7H11NO3
InChI KeyLOQFROBMBSKWQY-ZCFIWIBFSA-N
SMILESCCOC(=O)C[C@H](O)CC#N
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear, colourless to pale yellow
Refractive Index1.4460 - 1.4500 @20?C
Formliquid
Optical Rotation-4.5? ? 0.5?, neat
Assay (GC)> 97.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only