3,5-Difluoro-2-methoxybenzeneboronic acid, 97%, Thermo Scientific Chemicals
3,5-Difluoro-2-methoxybenzeneboronic acid, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

3,5-Difluoro-2-methoxybenzeneboronic acid, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
250 mg
1 g
Catalog number L19773.06
also known as L19773-06
Price (USD)/ Each
540.00
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
540.00
Add to cart
3,5-Difluoro-2-methoxybenzeneboronic acid, 97%, Thermo Scientific Chemicals
Catalog numberL19773.06
Price (USD)/ Each
540.00
-
Add to cart
Chemical Identifiers
CAS737000-76-9
IUPAC Name(3,5-difluoro-2-methoxyphenyl)boronic acid
Molecular FormulaC7H7BF2O3
InChI KeyJXQHRWOUVJRCSR-UHFFFAOYSA-N
SMILESCOC1=C(F)C=C(F)C=C1B(O)O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
FormCrystals or powder or crystalline powder
Assay (Aqueous acid-base Titration)≥96.0%
Melting Point (clear melt)109.0-119.0?C
Proton NMRConforms to structure
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only