3-Fluoropiperidine hydrochloride, 97%, Thermo Scientific Chemicals
3-Fluoropiperidine hydrochloride, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

3-Fluoropiperidine hydrochloride, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
250 mg
Catalog number L19776.03
also known as L19776-03
Price (USD)/ Each
297.65
Online exclusive
331.00 
Save 33.35 (10%)
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)/ Each
297.65
Online exclusive
331.00 
Save 33.35 (10%)
Add to cart
3-Fluoropiperidine hydrochloride, 97%, Thermo Scientific Chemicals
Catalog numberL19776.03
Price (USD)/ Each
297.65
Online exclusive
331.00 
Save 33.35 (10%)
-
Add to cart
Chemical Identifiers
CAS737000-77-0
IUPAC Name3-fluoropiperidine hydrochloride
Molecular FormulaC5H11ClFN
InChI KeyRDJUBLSLAULIAT-UHFFFAOYNA-N
SMILESCl.FC1CCCNC1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (Titration ex Chloride)≥96.0 to ≤104.0%
Appearance (Color)White to cream
FormPowder or lumps
Water Content (Karl Fischer Titration)≤1.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only