Bis(phenylacetyl) disulfide, 98%, Thermo Scientific Chemicals
Bis(phenylacetyl) disulfide, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Bis(phenylacetyl) disulfide, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Catalog number L19792.14
also known as L19792-14
Price (USD)/ Each
197.65
Online exclusive
219.00 
Save 21.35 (10%)
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
197.65
Online exclusive
219.00 
Save 21.35 (10%)
Add to cart
Bis(phenylacetyl) disulfide, 98%, Thermo Scientific Chemicals
Catalog numberL19792.14
Price (USD)/ Each
197.65
Online exclusive
219.00 
Save 21.35 (10%)
-
Add to cart
Chemical Identifiers
CAS15088-78-5
IUPAC Name2-phenyl-1-[(2-phenylacetyl)disulfanyl]ethan-1-one
Molecular FormulaC16H14O2S2
InChI KeyIXGZXXBJSZISOO-UHFFFAOYSA-N
SMILESO=C(CC1=CC=CC=C1)SSC(=O)CC1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale cream
FormCrystals or powder or crystalline powder
Assay (HPLC)≥97.5%
Identification (FTIR)Conforms
Melting Point (clear melt)56.0-62.0?C
Bis(phenylacetyl)disulfide as sulfur substitution agent in preparation of RNA oligonucleotides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Bis(phenylacetyl)disulfide as sulfur substitution agent in preparation of RNA oligonucleotides.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
RUO – Research Use Only

General References:

  1. Li Wu, Yuan Wang,; Junzhou Wu,; Cong Lv,; Jie Wang,; Xinjing Tang. Caged circular antisense oligonucleotides for photomodulation of RNA digestion and gene expression in cells. Nucl. Acids Res.. 2013, 41 (1), 677-686..
  2. Sulfur transfer reagent in oligonucleotide chemistry: Nucleosides Nucleotides, 18, 485 (1999).