Thermo Scientific Chemicals

2,6-Dichlorobenzeneboronic acid, 95%, Thermo Scientific Chemicals

Catalog number: L19816.03
1 g, Each
Thermo Scientific Chemicals

2,6-Dichlorobenzeneboronic acid, 95%, Thermo Scientific Chemicals

Catalog number: L19816.03
1 g, Each
Quantity
Catalog number: L19816.03
also known as L19816-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
73852-17-2
IUPAC Name
(2,6-dichlorophenyl)boronic acid
Molecular Formula
C6H5BCl2O2
InChI Key
CXDPUSMFYPQXCV-UHFFFAOYSA-N
SMILES
OB(O)C1=C(Cl)C=CC=C1Cl
Assay (Aqueous acid-base Titration)
≥94.0%
Appearance (Color)
White to cream
Melting Point (clear melt)
144.0-157.0?C
Form
Powder
Assay (HPLC)
≥94.0%

Description

Reagent used for palladium-catalyzed stille and suzuki-miyaura cross-couplings, palladium (pd)-catalytized reactions. Reagent used in preparation of common building blocks for pharmaceuticals and agrochemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Reagent used for palladium-catalyzed stille and suzuki-miyaura cross-couplings, palladium (pd)-catalytized reactions. Reagent used in preparation of common building blocks for pharmaceuticals and agrochemicals.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text