Thermo Scientific Chemicals

Furan-3-boronic acid, 97%, Thermo Scientific Chemicals

Catalog number: L19834.03
1 g, Each
Thermo Scientific Chemicals

Furan-3-boronic acid, 97%, Thermo Scientific Chemicals

Catalog number: L19834.03
1 g, Each
Quantity
Catalog number: L19834.03
also known as L19834-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
55552-70-0
IUPAC Name
(furan-3-yl)boronic acid
Molecular Formula
C4H5BO3
InChI Key
CYEFKCRAAGLNHW-UHFFFAOYSA-N
SMILES
OB(O)C1=COC=C1
Assay (HPLC)
≥96.0%
Appearance (Color)
White to cream or grey
Assay (Aqueous acid-base Titration)
≥96.0%
Form
Crystals or powder or crystalline powder
Proton NMR
Conforms to structure

Description

Furan-3-boronic acid use furan-3-boronic acid as our coupling partner in many reactions. A chitosan uricase (UOx)-poly (furan-3-boronic acid)(PFBA)-Pd nanoparticles (PdNPs)/plated Pd (Pd plate)/multiwalled carbon nanotubes (MWCNTs)/Au electrode was prepared for fabricating an amperometric biosensor and a biofuel cell (BFC) of uric acid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Furan-3-boronic acid use furan-3-boronic acid as our coupling partner in many reactions. A chitosan uricase (UOx)-poly (furan-3-boronic acid)(PFBA)-Pd nanoparticles (PdNPs)/plated Pd (Pd plate)/multiwalled carbon nanotubes (MWCNTs)/Au electrode was prepared for fabricating an amperometric biosensor and a biofuel cell (BFC) of uric acid.

Solubility
Soluble in methanol.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text