Thermo Scientific Chemicals

2-Amino-4-cyanopyridine, 97%, Thermo Scientific Chemicals

Catalog number: L19841.06
5 g, Each
Thermo Scientific Chemicals

2-Amino-4-cyanopyridine, 97%, Thermo Scientific Chemicals

Catalog number: L19841.06
5 g, Each
Quantity
Catalog number: L19841.06
also known as L19841-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
42182-27-4
IUPAC Name
2-aminopyridine-4-carbonitrile
Molecular Formula
C6H5N3
InChI Key
GEEAYLFEIFJFGP-UHFFFAOYSA-N
SMILES
NC1=CC(=CC=N1)C#N
Form
Crystals or powder or crystalline powder
Appearance (Color)
Cream to yellow to brown
Melting Point (clear melt)
146.0-156.0?C
Assay (GC)
≥96.0%

Description

2-Amino-4-cyanopyridine is used in the treatment of central nervous system (CNS) disorders and as a mGluR2 antagonist.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Amino-4-cyanopyridine is used in the treatment of central nervous system (CNS) disorders and as a mGluR2 antagonist.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text