2-Amino-4-cyanopyridine is used in the treatment of central nervous system (CNS) disorders and as a mGluR2 antagonist.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2-Amino-4-cyanopyridine is used in the treatment of central nervous system (CNS) disorders and as a mGluR2 antagonist.
Solubility
Slightly soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents.
RUO – Research Use Only
General References:
- Hasanvand, F.; Nasrollahi, N.; Vajed, A.; Amani, S. Synthesis, spectroscopy and characterization of four alkoxo-bridged dinuclear copper (II) complexes containing 2-amino-4-methylpyridine or 2-amino-4-cyanopyridine as the ligands. Malay. J. Chem. 2010, 12 27-32.
- Z. Tber.; M.-A. Hiebel.; H. Allouchi.; A. El Hakmaoui.; M. Akssira.; G. Guillaumet.; S. Berteina-Raboin. Metal free direct formation of various substituted pyrido[2',1':2,3]imidazo[4,5-c]isoquinolin-5-amines and their further functionalization. RSC Adv. 2015, 5 (44), 35201-35210.