2-Aminopyridine-4-carboxamide is used as pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2-Aminopyridine-4-carboxamide is used as pharmaceutical intermediate.
Solubility
Slightly soluble in water.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only
Christel J.Menet. Toward selective TYK2 inhibitors as therapeutic agents for the treatment of inflammatory diseases. Pharmaceutical Patent Analyst. 2014, 3,(4), 449-466.