Thermo Scientific Chemicals

2,4-Dibromobenzenesulfonyl chloride, 98%, Thermo Scientific Chemicals

Catalog number: L19887.06
5 g, Each
Thermo Scientific Chemicals

2,4-Dibromobenzenesulfonyl chloride, 98%, Thermo Scientific Chemicals

Catalog number: L19887.06
5 g, Each
Quantity
Catalog number: L19887.06
also known as L19887-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
72256-95-2
IUPAC Name
2,4-dibromobenzene-1-sulfonyl chloride
Molecular Formula
C6H3Br2ClO2S
InChI Key
OMOXIKAWVFDFNX-UHFFFAOYSA-N
SMILES
ClS(=O)(=O)C1=C(Br)C=C(Br)C=C1
Appearance (Color)
White to yellow
Melting Point (clear melt)
81.5-87.5?C
Form
Crystals or powder or crystalline powder or fused/lumpy solid
Assay (Titration ex Chloride)
≥97.5 to ≤102.5%

Description

2,4-Dibromobenzenesulfonyl chloride is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,4-Dibromobenzenesulfonyl chloride is used as pharmaceutical intermediate.

Solubility
Insoluble in water.

Notes
Moisture sensitive. Incompatible with oxidizing agents and bases.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text